(R)-3,3'-Bis(3,5-difluorophenyl)-[1,1'-binapthalene]-2,2'-diol ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

15-0066-250MG
101418-504EA 67.94 USD
101418-504
(R)-3,3'-Bis(3,5-difluorophenyl)-[1,1'-binapthalene]-2,2'-diol ≥98%
(R)-3,3'-Bis(3,5-difluorophenyl)-[1,1'-binapthalene]-2,2'-diol

Phosphine, SEGPHOS

Formula: C₃₂H₁₈F₄O₂
MW: 510.478 g/mol
Melting Pt: 126…128 °C
MDL Number: MFCD17019237
CAS Number: 1706439-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR