(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-4-Hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide(R)-(-)-1,1'-Binaphthalene-2,2'-diyl-phosphate

15-0052-1G 15-0052-5G
101418-482EA 72.18 USD
101418-482 101418-484
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥98%
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate
Formula: C₂₀H₁₃O₄P
MW: 348.289 g/mol
MDL Number: MFCD00010045
CAS Number: 39648-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR