Tetrakis(trimethylsilyl)silane ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: TMSS

14-7000-1G
100200-406EA 128.8 USD
100200-406
Tetrakis(trimethylsilyl)silane ≥98%
Tetrakis(trimethylsilyl)silane
Formula: C₁₂H₃₆Si₅
MW: 320.842 g/mol
MDL Number: MFCD00054859
CAS Number: 4098-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR