Bis(trimethylsilyl)methane ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
14-1500-5G
14-1500-1G
100208-308EA
65.11
USD
100208-308
100208-310
Bis(trimethylsilyl)methane ≥97%
Bis(trimethylsilyl)methane
Formula:
C₇H₂₀Si₂ Boiling Pt: 133…134 °C Melting Pt: –71 °C Density: 0,751 Flash Pt: 55 °F |
MDL Number:
MFCD00008275 CAS Number: 2117-28-4 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...