Phosphotungstic acid modified alumina metal scavenger (BASF MSA-FC Al-1)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
13-6300-50G
13-6300-10G
101324-540EA
533.6
USD
101324-540
101324-538
Phosphotungstic acid modified alumina metal scavenger (BASF MSA-FC Al-1)
Phosphotungstic acid modified alumina
MSA
- Color and Form: d50=159μm; white pwdr.
Sold in collaboration with BASF for research purposes only.BASF Metals Scavenging Agent Kit component
Density:
1.00 g/mL |
MDL Number:
MFCD07370730 CAS Number: 1344-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...