Aluminium trifluoromethanesulfonate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Aluminum tris(trifluoromethanesulphonate)
, Aluminum trifluoromethanesulphonate
, Aluminum triflate
, Aluminum trifluoromethanesulfonate
, Aluminum tris(trifluoromethanesulfonate)
13-1500-50G
13-1500-10G
100197-062EA
634.09
USD
100197-062
100200-846
Aluminium trifluoromethanesulfonate ≥99%
Aluminium trifluoromethanesulphonate
Metal Triflate
Formula:
C₃AlF₉O₉S₃ MW: 474.19 g/mol Melting Pt: Dec. |
MDL Number:
MFCD00143596 CAS Number: 74974-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...