Magnesium trifluoromethanesulfonate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Magnesium trifluoromethanesulfonate
, Magnesium trifluoromethanesulphonate
, Magnesium triflate
12-4000-5G
12-4000-25G
100196-032EA
63.69
USD
100196-032
100198-110
Magnesium trifluoromethanesulfonate ≥98%
Magnesium trifluoromethanesulphonate
Metal Triflate
Formula:
C₂F₆MgO₆S₂ MW: 322.45 g/mol |
MDL Number:
MFCD00042601 CAS Number: 60871-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...