3,3',5,5'-Tetramethyl-2,2'-biphenol ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-2,2'-diol

08-2043-500MG
100209-572EA 433.11 USD
100209-572
3,3',5,5'-Tetramethyl-2,2'-biphenol ≥99%
3,3',5,5'-Tetramethyl-2,2'-biphenol
Formula: C₁₆H₁₈O₂
MW: 242.3129 g/mol
Melting Pt: 134…136 °C
MDL Number: MFCD00854202
CAS Number: 26567-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR