(S)-(-)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(S)-5,5,6,6,7,7,8,8-Octahydro-1,1-bi-2-naphthol
, (S)-(-)-2,2'-Dihydroxy-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl
08-2036-1G
08-2036-250MG
100198-542EA
493.97
USD
100198-542
100209-570
(S)-(-)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol ≥99%
(S)-(-)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol
Formula:
C₂₀H₂₂O₂ MW: 294.39 g/mol |
MDL Number:
MFCD02093485 CAS Number: 65355-00-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...