(-)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(4R,5R)-(-)-2,2-Dimethyl-α,α,α',α'-tetraphenyl-1,3-dioxolane-4,5-dimethanol
, (-)-2,3-O-Isopropylidene-1,1,4,4-tetraphenyl-L-threitol
, (4R,5R)-2,2-Dimethyl-alpha,alpha,alpha',alpha'-tetraphenyldioxolane-4,5-dimethanol
, (R,R)-TADDOL
08-2008-500MG
08-2008-2G
100198-538EA
124.55
USD
100198-538
100206-256
(-)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane
(-)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane
TADDOL
Formula:
C₃₁H₃₀O₄ MW: 466.58 g/mol Melting Pt: 193…195 °C |
MDL Number:
MFCD00064467 CAS Number: 93379-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...