(R)-(+)-1,1'-Bi-2-naphthol ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(R)-BINOL
, (R)-(+)-1,1'-Binaphthyl-2,2'-Diol
, (R)-[1,1'-Binaphthalene]-2,2'-diol
08-1000-5G
08-1000-25G
08-1000-1G
100198-526EA
59.45
USD
100198-526
100198-528
100202-880
(R)-(+)-1,1'-Bi-2-naphthol ≥99%
(R)-(+)-1,1'-Bi-2-naphthol
BINOL
Formula:
C₂₀H₁₄O₂ MW: 286.33 g/mol Melting Pt: 208…211 °C |
MDL Number:
MFCD00004068 CAS Number: 18531-94-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...