(±)-6,6'-Dibromo-1,1'-bi-2-naphthol ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
6,6'-Dibromo[1,1'-binaphthalene]-2,2'-diol
, racemic-6,6'-Dibromo-1,1'-bi-2-naphthol
08-0604-5G
08-0604-1G
100208-280EA
343.94
USD
100208-280
100200-386
(±)-6,6'-Dibromo-1,1'-bi-2-naphthol ≥98%
(±)-6,6'-Dibromo-1,1'-bi-2-naphthol
Formula:
C₂₀H₁₂Br₂O₂ MW: 444.116 g/mol Melting Pt: 202…205 °C |
MDL Number:
MFCD00798290 CAS Number: 13185-00-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...