(R)-(+)-3,3'-Dibromo-1,1'-bi-2-naphthol ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

08-0600-100MG 08-0600-500MG
100200-382EA 134.46 USD
100200-382 100200-384
(R)-(+)-3,3'-Dibromo-1,1'-bi-2-naphthol ≥98%
(R)-(+)-3,3'-Dibromo-1,1'-bi-2-naphthol
Formula: C₂₀H₁₂Br₂O₂
Melting Pt: 254…258 °C
MDL Number: MFCD03093629
CAS Number: 111795-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR