Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dibenzo-18-crown-6

08-0500-50G 08-0500-10G
100198-510EA 351.02 USD
100198-510 100195-990
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) ≥98%
Dibenzo-18-crown-6 (Dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin (II) ionophore I)
Formula: C₂₀H₂₄O₆
MW: 360.41 g/mol
Melting Pt: 162…164 °C
MDL Number: MFCD00005098
CAS Number: 14187-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR