(S)-(-)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: (S)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl(S)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthalene

08-0171-250MG 08-0171-5G 08-0171-1G
100201-386EA 93.42 USD
100201-386 100201-388 100201-390
(S)-(-)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl ≥98%
(S)-(-)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl
Formula: C₂₄H₂₂O₄
MW: 374.44 g/mol
Melting Pt: 100…103 °C
MDL Number: MFCD03788935
CAS Number: 142128-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR