(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(S,S)-TsDPEN
, N-((1S,2S)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide
, N-((1S,2S)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulphonamide
, (1S,2S)-(+)-N-(4-toluenesulfonyl)-1,2-diphenylethylenediamine
, (S,S)-N-(p-Toluenesulfonyl)-1,2-diphenylethylenediamine
, (1S,2S)-(+)-N-(4-toluenesulphonyl)-1,2-diphenylethylenediamine
, (S,S)-N-(p-Toluenesulphonyl)-1,2-diphenylethylenediamine
CATHy Catalyst, DPEN
Formula:
C₂₁H₂₂N₂O₂S MW: 366.48 g/mol Melting Pt: 127…129 °C |
MDL Number:
MFCD03095684 CAS Number: 167316-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...