1,4,8,11-Tetraazacyclopentadecane ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1,4,8,12-Tetraazacyclopentadecane

07-1955-1G 07-1955-250MG
100198-502EA 84.92 USD
100198-502 100197-332
1,4,8,11-Tetraazacyclopentadecane ≥98%
1,4,8,11-Tetraazacyclopentadecane
Formula: C₁₁H₂₆N₄
MW: 214.35 g/mol
Melting Pt: 98…99 °C
MDL Number: MFCD00010049
CAS Number: 15439-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR