1-Methyl-3-propylimidazolium phosphate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

07-1725-5G
100209-540EA 418.95 USD
100209-540
1-Methyl-3-propylimidazolium phosphate ≥99%
1-Methyl-3-propylimidazolium phosphate

Ionic Liquids

MDL Number: MFCD03788932
CAS Number: 817575-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR