1-Ethyl-3-methylimidazolium bis(pentafluoroethylsulfonyl)imide ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: EMIBeti

07-0578-2G
100209-536EA 638.34 USD
100209-536
1-Ethyl-3-methylimidazolium bis(pentafluoroethylsulfonyl)imide ≥99%
1-Ethyl-3-methylimidazolium bis(pentafluoroethylsulfonyl)imide

Ionic Liquids

Density: 1.57
MDL Number: MFCD03788926
CAS Number: 216299-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR