1,3-Dimesityl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate ≥95%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
1,3-Bis(2,4,6-trimethylphenyl)imidazolinium tetrafluoroborate
, 1,3-Bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium tetrafluoroborate
, SIMes-HBF4
07-0302-1G
07-0302-5G
100195-960EA
489.72
USD
100195-960
100195-958
1,3-Dimesityl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate ≥95%
1,3-Dimesityl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate
NHC
Formula:
C₂₁H₂₇BF₄N₂ MW: 394.26 g/mol |
MDL Number:
MFCD02684542 CAS Number: 245679-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...