2,6-Bis[1-(2,6-di-i-propylphenylimino)ethyl]pyridine ≥98%
Supplier: STREM CHEMICALS INC MS
Synonyms:
2,6-Bis-[1-(2,6-diisopropylphenylimino)ethyl]pyridine
07-0289-500MG
07-0289-2G
100200-236EA
226.46
USD
100200-236
100205-912
2,6-Bis[1-(2,6-di-i-propylphenylimino)ethyl]pyridine ≥98%
2,6-Bis[1-(2,6-di-i-propylphenylimino)ethyl]pyridine
Pincer Ligands and Complexes
Formula:
C₃₃H₄₃N₃ MW: 481.7146 g/mol |
MDL Number:
MFCD01862436 CAS Number: 204203-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...