(S,S)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline) ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(S,S)-2,2'-(Dimethylmethylene)bis(4-phenyl-2-oxazoline)
, (4S,4S)-2,2-(Propane-2,2-diyl)bis(4-phenyl-4,5-dihydrooxazole)
, (-)-2,2-Bis[(4S)-4-phenyl-2-oxazolin-2-yl]propane
, (S,S)-2,2-Bis(4-phenyl-2-oxazolin-2-yl)propane
07-0275-1G
07-0275-250MG
100197-316EA
164.18
USD
100197-316
100207-328
(S,S)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline) ≥98%
(S,S)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline)
Formula:
C₂₁H₂₂N₂O₂ MW: 334.42 g/mol |
MDL Number:
MFCD00192245 CAS Number: 131457-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...