Poly(styrene-divinylbenzene), 20% cross-linked

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Poly(styrene-co-divinylbenzene)Styrene-divinylbenzene resinSDVB resinStyrene-divinylbenzene copolymer

06-2000-250G 06-2000-50G
100204-790EA 41.05 USD
100204-790 100199-312
Poly(styrene-divinylbenzene), 20% cross-linked
Poly(styrene-divinylbenzene)
Formula: [-CH₂CH(C₆H₅)-]x[-CH₂CH[C₆H₄(CHCH₂)]-]y
MW: 234.342 g/mol
MDL Number: MFCD00146442
CAS Number: 9003-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR