i-Propylisocyanide ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
06-1850-5G
06-1850-1G
100202-846EA
1851.32
USD
100202-846
100202-844
i-Propylisocyanide ≥97%
i-Propylisocyanide
Boiling Pt:
82…83 °C Density: 0,7596 |
MDL Number:
MFCD00015503 CAS Number: 598-45-8 UN: 1992 ADR: 3,II |