Phosphotungstic acid modified activated carbon metal scavenger (BASF MSA-FC C-1)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
06-0805-10G
06-0805-50G
101324-504EA
826.58
USD
101324-504
101324-502
Phosphotungstic acid modified activated carbon metal scavenger (BASF MSA-FC C-1)
Phosphotungstic acid modified activated carbon
MSA
- Color and Form: d50=27μm; black pwdr.
Sold in collaboration with BASF for research purposes only.
Density:
0.60 g/mL |
MDL Number:
MFCD07370731 CAS Number: 7440-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...