tert-Butyl isocyanide ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
t-Butylisocyanide
06-0120-5G
06-0120-1G
100202-350EA
383.57
USD
100202-350
100197-302
tert-Butyl isocyanide ≥98%
tert-Butyl isocyanide
Formula:
C₅H₉N MW: 83.13 g/mol Boiling Pt: 90…92 °C Density: 0.735 Flash Pt: 28 °F |
MDL Number:
MFCD00000002 CAS Number: 7188-38-7 UN: 1992 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...