n-Butyl isocyanide ≥97%

Danger

06-0115-5G 06-0115-1G
100195-952EA 175.51 USD
100195-952 100206-074
n-Butyl isocyanide ≥97%
n-Butyl isocyanide
Formula: C₅H₉N
Density: 0,795
UN: 1992
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR