Tris(trimethylsilyl)borate ≥97%

Warning

Synonyms: Tris(trimethylsiloxy)borate

05-2000-50G 05-2000-10G
100195-950EA 150 USD
100195-950 100209-504
Tris(trimethylsilyl)borate ≥97%
Tris(trimethylsilyl) borate
Formula: C₉H₂₇BO₃Si₃
MW: 278.376 g/mol
Boiling Pt: 184 °C
Melting Pt: –35 °C
Density: 0,828
Flash Pt: 106 °F
MDL Number: MFCD00051588
CAS Number: 4325-85-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR