4-(Methylthio)phenylboronic acid ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
4-Thioanisoleboronic acid
, 4-(Methylthio)benzeneboronic acid
05-1010-5G
05-1010-1G
100200-826EA
188.25
USD
100200-826
100195-942
4-(Methylthio)phenylboronic acid ≥97%
4-(Methylthio)phenylboronic acid
Boronic Acid and Ester
Formula:
C₇H₉BO₂S MW: 168.02 g/mol Melting Pt: 210…214 °C |
MDL Number:
MFCD00093410 CAS Number: 98546-51-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...