Bis(pinacolato)diboron ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane)
, 4,4,5,5-tetramethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane
05-0030-500MG
05-0030-2G
100200-370EA
38.22
USD
100200-370
100203-688
Bis(pinacolato)diboron ≥99%
Bis(pinacolato)diboron
Boronic Acid and Ester
Formula:
C₁₂H₂₄B₂O₄ MW: 253.94 g/mol Melting Pt: 144…145 °C Density: 0,98 |
MDL Number:
MFCD00799570 CAS Number: 73183-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...