Lithium hexafluoroantimonate ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

03-1200-25G 03-1200-5G
100206-178EA 621.35 USD
100206-178 100206-180
Lithium hexafluoroantimonate ≥97%
Lithium hexafluoroantimonate
Formula: F₆LiSb
MW: 242.68
MDL Number: MFCD00042319
CAS Number: 18424-17-4
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR