Human Recombinant Signal Transducer and Activator of Transcription 6
Supplier: Bon Opus Biosciences
CG37-50UG CG37-1MG CG37-10UG CG37-500UG
102865-340EA 497.54 USD
102865-340 102865-334 102865-336 102865-338
Human Recombinant Signal Transducer and Activator of Transcription 6
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human STAT6 is produced by our E.coli expression system and the target gene encoding Ser627-Ser837 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.