Recombinant Human Ube2C
Supplier: Bon Opus Biosciences
CF41-1MG CF41-50UG CF41-10UG CF41-500UG
102864-752EA 2898 USD
102864-752 102864-750 102864-756 102864-754
Recombinant Human Ube2C
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human ubiquitin-conjugating enzyme E2 C is produced by our E.coli expression system and the target gene encoding Met1-Pro179 is expressed with a 6His, T7 tag at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx.
Packaging: Shipped domestic on polar packs in a 9×6×7" box, or shipped internationally on dry ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.