Human Recombinant Interferon Regulatory Factor 5
Supplier: Bon Opus Biosciences
CF18-50UG CF18-10UG CF18-1MG CF18-500UG
102864-606EA 170.15 USD
102864-606 102864-608 102864-610 102864-612
Human Recombinant Interferon Regulatory Factor 5
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Interferon Regulatory Factor 5 is produced by our E.coli expression system and the target gene encoding Met1-Gln498 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.