Human Recombinant C-C Motif Chemokine 24
Supplier: Bon Opus Biosciences
CC43-500UG CC43-10UG CC43-50UG CC43-1MG
102497-452EA 322 USD
102497-452 102497-450 102497-448 102497-446
Human Recombinant C-C Motif Chemokine 24
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human C-C Motif Chemokine 24 is produced by our Mammalian expression system and the target gene encoding Val27-Cys119 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.