Human Recombinant Tumor Necrosis Factor Receptor Superfamily Member 21
Supplier: Bon Opus Biosciences
C327-50UG C327-10UG C327-1MG C327-500UG
102861-382EA 45.23 USD
102861-382 102861-384 102861-386 102861-388
Human Recombinant Tumor Necrosis Factor Receptor Superfamily Member 21
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Death Receptor 6 is produced by our Mammalian expression system and the target gene encoding Gln42-Leu350 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.