    Sort by:
  • Search Within Results
P529
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   P529, A potent PI3K/Akt/mTOR inhibitor, Synonyms: 3-(4-methoxybenzyloxy)-8-(1-hydroxyethyl)-2-methoxy-6H-benzo[c]chromen-6-one ; Palomid 529, CAS Number: 914913-88-5, Purity: ≥98% by HPLC, 5 mg