    Sort by:
  • Search Within Results
PluriSIn 1
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   PluriSIn 1, A selective steroyl-CoA desaturase 1 (SCD1) inhibitor, Alternate Name/Synonyms: 4-Pyridinecarboxylic acid, 2-phenylhydrazide; NSC-14613, CAS Number: 91396-88-2, Purity: ≥98% by HPLC, Molecular Formula: C12H11N3O, 10 mg