    Sort by:
  • Search Within Results
R5C3, 5-Methyl-N-(2-phenoxybenzoyl)-1-[(phenylmethoxy)carbonyl]-D/L-tryptophan
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   A MDM2 inhibitor and 2-phenoxybenzoyl-tryptophan derivative. Reported as a novel probe for high throughput fluorescence polarization binding assay