    Sort by:
You Searched For:
NSC 405020
1  results were found
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   NSC-405020 is a membrane type-I matrix metalloproteinase (MT1-MMP) inhibitor that specifically targets the hemopexin (PEX) domain rather than the cata...