    Sort by:
  • Search Within Results
Ivachtin
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   Ivachtin, a Caspase-3 inhibitor, Exhibits antiapoptotic activity in human Jurkat T cells, Purity: ≥98% by HPLC, Molecular Formula: C20H21N3O7S, Synonyms: 2-(4-methyl-8-morpholin-4-ylsulfonyl-1,3-dioxopyrrolo[3,4-c]quinolin-2-yl)ethyl acetate, Storage: -20 Degree C, Size: 5mg