    Sort by:
  • Search Within Results
IWP-3
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   IWP-3, An inhibitor of Wnt production, Synonyms: Inhibitor of Wnt Production-3; 2-​[[3-​(4-​fluorophenyl)-​3,​4,​6,​7-​tetrahydro-​4-​oxothieno[3,​2-​ d]pyrimidin-​2-​yl]thio]-​N-​(6-​methyl-​2-​benzothiazolyl)-​acetamide, CAS Number: 687561-60-0, Purity: ≥98%, 1 mg