    Sort by:
  • Search Within Results
Gliotoxin
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   Immunomodulating mycotoxin which acts by blocking membrane thiol groups. Inhibits the chymotrypsin-like activity of the 20S proteasome in a non-compet...