    Sort by:
  • Search Within Results
LDN-57444
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   A selective UCHL1 inhibitor, Synonyms: 3-(O-acetyloxime), 5-chloro-1-[(2,5-dichlorophenyl)methyl]-1H-Indole-2,3-dione, C30, LDN 57444, CAS Number: 668467-91-2, Molecular Formula: C1₇H11Cl₃N2O₃, Purity: ≥98% by TLC, 5mg.