    Sort by:
  • Search Within Results
Diethylamine NONOate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (80051-400)
Supplier:  MilliporeSigma
Description:   Reagent that releases nitric oxide (NO) in aqueous solutions at controlled rates. Half-life (t1/2 ) of NO release = 16 min (PBS, pH 7.4, 22[degree]C). CAS: 56329-27-2. Purity: >= 98% by elemental analysis. Soluble in water. FW: 206.3. 10mg white crystalline solid.