    Sort by:
Tryprostatin A
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   Diketopiperazine. Arrests cell cycle progression at the G2/M phase. Inhibitor of MAP-dependent microtubule assembly. Inhibits the ABC-transporter brea...