    Sort by:
  • Search Within Results
VX-702
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   A p38 MAP kinase inhibitor, Synonyms: 2-(2,4-Difluorophenyl)-6-(1-(2,6-difluorophenyl)ureido)nicotinamide, CAS Number: 745833-23-2, Purity: ≥99% by High-performance liquid chromatography, Molecular Formula: C19H12F₄N₄O2, 5mg.