    Sort by:
  • Search Within Results
VO-OHpic trihydrate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   VO-OHpic, Trihydrate, A PTP and PTEN inhibitor, Synonyms: [VO(Hhpic-O,O) (Hhpic-O,N)(H2O)]. 3H2O, CAS Number: 476310-60-8, Purity: ≥98% by V-NMR, 5 mg