You Searched For: Coumermycin A1
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (89158-902)
Supplier: Enzo Life Sciences
Description: Inhibits supercoiling activity of gyrase by binding to the gyrase B domain – but more importantly is a novel cell permeable dimerizing ligand fo...