    Sort by:
  • Search Within Results
bpV(phen)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (10188-150)
Supplier:  BioVision
Description:   A PTP and PTEN inhibitor, Synonyms: Potassium bisperoxo(1,10-phenanthroline)oxovanadate (V), Purity: ≥98% by Nuclear Magnetic Resonance, CAS Number: 42494-73-5, Molecular Formula: K[VO(O2)2C12H8N2].3H2O, 5mg.